Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607559 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MGRN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MGRN1 - mahogunin ring finger 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142289.2 | Intron | NP_001135761.2 | ||||
NM_001142290.2 | Intron | NP_001135762.1 | ||||
NM_001142291.2 | Intron | NP_001135763.2 | ||||
NM_015246.3 | Intron | NP_056061.1 | ||||
XM_005255217.1 | Intron | XP_005255274.1 | ||||
XM_005255218.1 | Intron | XP_005255275.1 | ||||
XM_005255219.3 | Intron | XP_005255276.1 | ||||
XM_005255220.3 | Intron | XP_005255277.1 | ||||
XM_005255221.3 | Intron | XP_005255278.1 | ||||
XM_011522441.1 | Intron | XP_011520743.1 | ||||
XM_011522442.2 | Intron | XP_011520744.1 | ||||
XM_017023091.1 | Intron | XP_016878580.1 | ||||
XM_017023092.1 | Intron | XP_016878581.1 | ||||
XM_017023093.1 | Intron | XP_016878582.1 |
UBALD1 - UBA like domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |