Search
Search

Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FADS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
| FADS3 - fatty acid desaturase 3 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| RAB3IL1 - RAB3A interacting protein like 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001271686.1 | 2149 | UTR 3 | NP_001258615.1 | |||
| NM_013401.3 | 2149 | UTR 3 | NP_037533.2 | |||
| XM_005274140.4 | 2149 | UTR 3 | XP_005274197.1 | |||
| XM_006718647.3 | 2149 | UTR 3 | XP_006718710.1 | |||
| XM_011545196.2 | 2149 | Intron | XP_011543498.1 | |||
| XM_011545197.2 | 2149 | Intron | XP_011543499.1 | |||
| XM_011545198.2 | 2149 | Intron | XP_011543500.1 | |||
| XM_011545199.2 | 2149 | Intron | XP_011543501.1 | |||
| XM_011545200.2 | 2149 | Intron | XP_011543502.1 | |||
| XM_011545201.2 | 2149 | Intron | XP_011543503.1 | |||
Set Membership: |
|