Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
GGGACTGAGTGTTTATGGTAGAACT[A/G]TAGGCGGGACAGGAGCTTTGTGACG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611542 | ||||||||||||||||||||
Literature Links: |
ARSB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARSB - arylsulfatase B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000046.3 | 2006 | Nonsense Mutation | CAG,TAG | Q,* 503 | NP_000037.2 | |
NM_198709.2 | 2006 | Intron | NP_942002.1 | |||
XM_011543390.1 | 2006 | Nonsense Mutation | CAG,TAG | Q,* 503 | XP_011541692.1 | |
XM_011543391.2 | 2006 | Intron | XP_011541693.1 | |||
XM_011543392.2 | 2006 | Intron | XP_011541694.1 | |||
XM_011543393.1 | 2006 | Intron | XP_011541695.1 | |||
XM_017009471.1 | 2006 | Intron | XP_016864960.1 |