Search
Search

AKR7A3
LOC100506730AGGTATTCCCAAAGAAGCGGCCCAC[G/A]GGCTGTTTCCCATTCTTGTCCTCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
30 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKR7A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| AKR7A3 - aldo-keto reductase family 7 member A3 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_012067.2 | 468 | Silent Mutation | CCC,CCT | P,P 219 | NP_036199.2 | |
| XM_011541046.2 | 468 | Silent Mutation | CCC,CCT | P,P 219 | XP_011539348.1 | |
| XM_017000714.1 | 468 | Silent Mutation | CCC,CCT | P,P 148 | XP_016856203.1 | |
| LOC100506730 - uncharacterized LOC100506730 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||