Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614168 | ||||||||||||||||||||||||||||||||
Literature Links: |
PCK1 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global - Not Available | Caucasian
|
CEPH (CEU)
|
||||||
EAS - Not Available | African American
|
YRI (Yoruba)
|
||||||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR - Not Available | ||||||||
AMR - Not Available |
PCK1 - phosphoenolpyruvate carboxykinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002591.3 | Intron | NP_002582.3 | ||||
XM_011528839.1 | Intron | XP_011527141.1 |
Set Membership: |
HapMap Validated |