Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › C___2535786_10
          See other DBH GT Assays ›
          SNP ID:
          rs1611115
          Gene
          DBH
          Gene Name
          dopamine beta-hydroxylase
          Set Membership:
          > HapMap
          Chromosome Location:
          Chr.9: 133635393 - 133635393 on Build GRCh38
          Polymorphism:
          C/T, Transition substitution
          Context Sequence [VIC/FAM]:

          AAGGCAGCTGCCCTCAGTCTACTTG[C/T]GGGAGAGGACAGGAGGGAGAGGTGC

          Assay ID C___2535786_10
          Size
          Availability Made To Order
          Catalog # 4351379
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details



          Species:

          Human

          dbSNP Submissions:

          NA

          Phenotype:

          MIM: 609312

          Literature Links:

          DBH PubMed Links

          Allele Nomenclature:

          Minor Allele Frequency:

          1000Genome Applied Biosystems® HapMap
          Global
          T (0.21)
          (0.79)
          Caucasian - Not Available CEPH (CEU)
          T (0.19)
          (0.81)
          EAS
          T (0.19)
          (0.81)
          African American - Not Available YRI (Yoruba)
          T (0.17)
          (0.83)
          SAS
          T (0.20)
          (0.80)
          Chinese - Not Available JPT (Japanese)
          T (0.18)
          (0.82)
          AFR
          T (0.19)
          (0.81)
          Japanese - Not Available CHB (Han Chinese)
          T (0.22)
          (0.78)
          EUR
          T (0.21)
          (0.79)
          AMR
          T (0.32)
          (0.68)
          DBH - dopamine beta-hydroxylase
          Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
          NM_000787.3 Intron NP_000778.3

          Back To Top

          More Information


          Set Membership:

          HapMap

          Panther Classification:

          Molecular Function -

          hydroxylase oxidoreductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          cytokine production
          blood vessel remodeling
          response to amphetamine
          leukocyte mediated immunity
          chemical synaptic transmission
          memory
          locomotory behavior
          visual learning
          regulation of cell proliferation
          homoiothermy
          dopamine catabolic process
          norepinephrine biosynthetic process
          catecholamine biosynthetic process
          glucose homeostasis
          fear response
          maternal behavior
          positive regulation of vasoconstriction
          behavioral response to ethanol
          response to pain
          leukocyte migration
          oxidation-reduction process
          regulation of extrinsic apoptotic signaling pathway
          catalytic activity
          dopamine beta-monooxygenase activity
          copper ion binding
          L-ascorbic acid binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-knt4q:80/100.66.75.14:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0