Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137161 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GABRR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GABRR1 - gamma-aminobutyric acid type A receptor rho1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256703.1 | 1589 | Silent Mutation | GCA,GCG | A,A 378 | NP_001243632.1 | |
NM_001256704.1 | 1589 | Silent Mutation | GCA,GCG | A,A 308 | NP_001243633.1 | |
NM_001267582.1 | 1589 | Silent Mutation | GCA,GCG | A,A 308 | NP_001254511.1 | |
NM_002042.4 | 1589 | Silent Mutation | GCA,GCG | A,A 395 | NP_002033.2 | |
XM_017010689.1 | 1589 | Silent Mutation | GCA,GCG | A,A 308 | XP_016866178.1 |
Set Membership: |
HapMap |