Search
Search

Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MGAT4C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||||||||
|---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
| MGAT4C - MGAT4 family member C | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_013244.3 | Intron | NP_037376.2 | ||||
| XM_005268776.4 | Intron | XP_005268833.1 | ||||
| XM_005268779.3 | Intron | XP_005268836.1 | ||||
| XM_011538150.2 | Intron | XP_011536452.1 | ||||
| XM_011538151.2 | Intron | XP_011536453.1 | ||||
| XM_011538156.2 | Intron | XP_011536458.1 | ||||
| XM_011538157.2 | Intron | XP_011536459.1 | ||||
| XM_017019143.1 | Intron | XP_016874632.1 | ||||
| XM_017019144.1 | Intron | XP_016874633.1 | ||||
| XM_017019145.1 | Intron | XP_016874634.1 | ||||
| XM_017019146.1 | Intron | XP_016874635.1 | ||||
| XM_017019147.1 | Intron | XP_016874636.1 | ||||
| XM_017019148.1 | Intron | XP_016874637.1 | ||||
| XM_017019149.1 | Intron | XP_016874638.1 | ||||
| XM_017019150.1 | Intron | XP_016874639.1 | ||||
Set Membership: |
|