Search
Search

Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C19orf45 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||||||||
|---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
| C19orf45 - chromosome 19 open reading frame 45 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_198534.2 | Intron | NP_940936.2 | ||||
| XM_005272473.3 | Intron | XP_005272530.1 | ||||
| XM_011527993.1 | Intron | XP_011526295.1 | ||||
| XM_011527995.2 | Intron | XP_011526297.1 | ||||
| PEX11G - peroxisomal biogenesis factor 11 gamma | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||