Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604814 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LILRB5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LILRB5 - leukocyte immunoglobulin like receptor B5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081442.2 | Intron | NP_001074911.1 | ||||
NM_001081443.2 | Intron | NP_001074912.1 | ||||
NM_001304457.1 | Intron | NP_001291386.1 | ||||
NM_006840.4 | Intron | NP_006831.1 | ||||
XM_011526359.2 | Intron | XP_011524661.1 | ||||
XM_011526360.2 | Intron | XP_011524662.1 | ||||
XM_011526361.2 | Intron | XP_011524663.1 | ||||
XM_011526362.2 | Intron | XP_011524664.1 |
Set Membership: |
HapMap |