Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606969 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DBIL5P PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
DBIL5P - diazepam binding inhibitor-like 5, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GEMIN4 - gem nuclear organelle associated protein 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GLOD4 - glyoxalase domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016080.3 | Intron | NP_057164.3 | ||||
XM_006721540.2 | Intron | XP_006721603.1 | ||||
XM_017024711.1 | Intron | XP_016880200.1 | ||||
XM_017024712.1 | Intron | XP_016880201.1 |