Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
METTL6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
METTL6 - methyltransferase like 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301790.1 | Intron | NP_001288719.1 | ||||
NM_001301791.1 | Intron | NP_001288720.1 | ||||
NM_001301792.1 | Intron | NP_001288721.1 | ||||
NM_152396.3 | Intron | NP_689609.2 | ||||
XM_005264867.3 | Intron | XP_005264924.1 | ||||
XM_005264870.4 | Intron | XP_005264927.1 | ||||
XM_006712970.3 | Intron | XP_006713033.1 | ||||
XM_006712972.3 | Intron | XP_006713035.1 | ||||
XM_011533357.2 | Intron | XP_011531659.1 | ||||
XM_017005718.1 | Intron | XP_016861207.1 | ||||
XM_017005719.1 | Intron | XP_016861208.1 | ||||
XM_017005720.1 | Intron | XP_016861209.1 | ||||
XM_017005721.1 | Intron | XP_016861210.1 | ||||
XM_017005722.1 | Intron | XP_016861211.1 | ||||
XM_017005723.1 | Intron | XP_016861212.1 | ||||
XM_017005724.1 | Intron | XP_016861213.1 |
Set Membership: |
HapMap |