Search
Search

Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCL2L14 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||||||||
|---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
| BCL2L14 - BCL2 like 14 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_030766.1 | 681 | Silent Mutation | TAC,TAT | Y,Y 162 | NP_110393.1 | |
| NM_138722.1 | 681 | Silent Mutation | TAC,TAT | Y,Y 162 | NP_620048.1 | |
| NM_138723.1 | 681 | Silent Mutation | TAC,TAT | Y,Y 162 | NP_620049.1 | |
| XM_006719151.3 | 681 | Silent Mutation | TAC,TAT | Y,Y 162 | XP_006719214.1 | |
| XM_011520848.1 | 681 | Silent Mutation | TAC,TAT | Y,Y 162 | XP_011519150.1 | |
| XM_011520849.1 | 681 | Silent Mutation | TAC,TAT | Y,Y 162 | XP_011519151.1 | |
| XM_017019963.1 | 681 | Silent Mutation | TAC,TAT | Y,Y 162 | XP_016875452.1 | |