Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
AGGATCTGGCCTGAGACTCTCCTGC[C/T]GAGGTTCAGGAACGAGCCAGGTGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603506 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRP5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LRP5 - LDL receptor related protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291902.1 | Intron | NP_001278831.1 | ||||
NM_002335.3 | Intron | NP_002326.2 | ||||
XM_005273994.2 | Intron | XP_005274051.1 | ||||
XM_011545029.1 | Intron | XP_011543331.1 | ||||
XM_011545030.1 | Intron | XP_011543332.1 | ||||
XM_011545031.1 | Intron | XP_011543333.1 | ||||
XM_017017735.1 | Intron | XP_016873224.1 | ||||
XM_017017736.1 | Intron | XP_016873225.1 |
Set Membership: |
HapMap |