Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606557 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCL11A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
BCL11A - B-cell CLL/lymphoma 11A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018014.3 | 1434 | UTR 3 | NP_060484.2 | |||
NM_022893.3 | 1434 | Intron | NP_075044.2 | |||
NM_138559.1 | 1434 | UTR 3 | NP_612569.1 | |||
XM_011532909.1 | 1434 | Intron | XP_011531211.1 | |||
XM_011532910.1 | 1434 | Intron | XP_011531212.1 | |||
XM_011532912.1 | 1434 | Intron | XP_011531214.1 | |||
XM_017004333.1 | 1434 | Intron | XP_016859822.1 | |||
XM_017004334.1 | 1434 | Intron | XP_016859823.1 | |||
XM_017004335.1 | 1434 | Intron | XP_016859824.1 | |||
XM_017004336.1 | 1434 | Intron | XP_016859825.1 | |||
XM_017004337.1 | 1434 | Intron | XP_016859826.1 | |||
XM_017004338.1 | 1434 | Intron | XP_016859827.1 |
Set Membership: |
HapMap Validated |