Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142980 MIM: 142981 MIM: 142985 MIM: 611576 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HOXD-AS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
HOXD-AS2 - HOXD cluster antisense RNA 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXD3 - homeobox D3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006898.4 | Intron | NP_008829.3 | ||||
XM_005246509.3 | Intron | XP_005246566.1 | ||||
XM_005246510.4 | Intron | XP_005246567.1 | ||||
XM_005246511.3 | Intron | XP_005246568.1 | ||||
XM_005246513.4 | Intron | XP_005246570.1 | ||||
XM_006712477.2 | Intron | XP_006712540.1 | ||||
XM_011511065.2 | Intron | XP_011509367.1 | ||||
XM_011511066.2 | Intron | XP_011509368.1 |
HOXD4 - homeobox D4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXD8 - homeobox D8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR10B - microRNA 10b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |