Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611636 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NAALAD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NAALAD2 - N-acetylated alpha-linked acidic dipeptidase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300930.1 | Intron | NP_001287859.1 | ||||
NM_005467.3 | Intron | NP_005458.1 | ||||
XM_017017043.1 | Intron | XP_016872532.1 | ||||
XM_017017044.1 | Intron | XP_016872533.1 | ||||
XM_017017045.1 | Intron | XP_016872534.1 | ||||
XM_017017046.1 | Intron | XP_016872535.1 |
Set Membership: |
Validated |