Search
Search

Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
C7orf34 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| C7orf34 - chromosome 7 open reading frame 34 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_178829.4 | 1473 | Intron | NP_849151.2 | |||
| KEL - Kell blood group, metallo-endopeptidase | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_000420.2 | 1473 | Missense Mutation | AAC,CAC | N,H 698 | NP_000411.1 | |
| XM_005249993.2 | 1473 | Missense Mutation | AAC,CAC | N,H 710 | XP_005250050.1 | |
| XM_005249994.3 | 1473 | Missense Mutation | AAC,CAC | N,H 389 | XP_005250051.1 | |
| TRPV5 - transient receptor potential cation channel subfamily V member 5 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||