Search
Search

Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM117A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
| FAM117A - family with sequence similarity 117 member A | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| SLC35B1 - solute carrier family 35 member B1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001278784.1 | 999 | UTR 3 | NP_001265713.1 | |||
| NM_005827.2 | 999 | UTR 3 | NP_005818.2 | |||
| XM_005256929.3 | 999 | UTR 3 | XP_005256986.1 | |||
| XM_006721632.3 | 999 | UTR 3 | XP_006721695.1 | |||
| XM_011524179.2 | 999 | UTR 3 | XP_011522481.1 | |||
| XM_011524180.2 | 999 | UTR 3 | XP_011522482.1 | |||