Search
Search

Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
38 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNST PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
| CNST - consortin, connexin sorting protein | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001139459.1 | 4274 | Intron | NP_001132931.1 | |||
| NM_152609.2 | 4274 | UTR 3 | NP_689822.2 | |||
| XM_005273081.3 | 4274 | UTR 3 | XP_005273138.2 | |||
| XM_005273083.4 | 4274 | UTR 3 | XP_005273140.1 | |||
| XM_011544110.2 | 4274 | UTR 3 | XP_011542412.1 | |||
| XM_011544111.1 | 4274 | UTR 3 | XP_011542413.1 | |||
| XM_011544112.1 | 4274 | UTR 3 | XP_011542414.1 | |||
| XM_011544113.1 | 4274 | UTR 3 | XP_011542415.1 | |||
| XM_011544114.2 | 4274 | UTR 3 | XP_011542416.1 | |||
| XM_017000487.1 | 4274 | UTR 3 | XP_016855976.1 | |||
| XM_017000488.1 | 4274 | UTR 3 | XP_016855977.1 | |||
| XM_017000489.1 | 4274 | UTR 3 | XP_016855978.1 | |||