Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATATATCCAGTCATTTGAGTCTT[C/G]CAGTTAGAACATTCAACTTGACTAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609211 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC104968399 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC104968399 - uncharacterized LOC104968399 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MYL12A - myosin light chain 12A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MYL12B - myosin light chain 12B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144944.1 | Intron | NP_001138416.1 | ||||
NM_001144945.1 | Intron | NP_001138417.1 | ||||
NM_033546.3 | Intron | NP_291024.1 |
Set Membership: |
HapMap |