Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MS4A15 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MS4A15 - membrane spanning 4-domains A15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098835.1 | 360 | Missense Mutation | CGG,CTG | R,L 47 | NP_001092305.1 | |
NM_001278242.1 | 360 | Missense Mutation | CGG,CTG | R,L 47 | NP_001265171.1 | |
NM_152717.2 | 360 | Intron | NP_689930.1 | |||
XM_006718459.3 | 360 | Intron | XP_006718522.1 | |||
XM_011544812.2 | 360 | Missense Mutation | CGG,CTG | R,L 47 | XP_011543114.1 | |
XM_011544814.1 | 360 | UTR 5 | XP_011543116.1 |
Set Membership: |
HapMap |