Search
Search

Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACVR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||||||||
|---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
| ACVR1 - activin A receptor type 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001105.4 | 2598 | UTR 3 | NP_001096.1 | |||
| NM_001111067.2 | 2598 | UTR 3 | NP_001104537.1 | |||
| XM_005246939.3 | 2598 | UTR 3 | XP_005246996.1 | |||
| XM_005246940.3 | 2598 | UTR 3 | XP_005246997.1 | |||
| XM_006712825.3 | 2598 | UTR 3 | XP_006712888.1 | |||
| XM_011512106.2 | 2598 | UTR 3 | XP_011510408.1 | |||
| XM_011512107.2 | 2598 | UTR 3 | XP_011510409.1 | |||
| XM_011512108.2 | 2598 | UTR 3 | XP_011510410.1 | |||
Set Membership: |
|