Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605790 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
STK31 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
STK31 - serine/threonine kinase 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001260504.1 | Intron | NP_001247433.1 | ||||
NM_001260505.1 | Intron | NP_001247434.1 | ||||
NM_031414.4 | Intron | NP_113602.2 | ||||
NM_032944.3 | Intron | NP_116562.2 | ||||
XM_006715758.3 | Intron | XP_006715821.1 | ||||
XM_011515449.2 | Intron | XP_011513751.2 | ||||
XM_011515450.1 | Intron | XP_011513752.1 | ||||
XM_011515452.2 | Intron | XP_011513754.2 |