Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › C___7817765_C0
          See other CYP2B6 GT Assays ›
          SNP ID:
          rs3745274
          Gene
          CYP2B6
          Gene Name
          cytochrome P450 family 2 subfamily B member 6
          Set Membership:
          > HapMap > DME > Validated > Inventoried
          Chromosome Location:
          Chr.19: 41006936 - 41006936 on Build GRCh38
          Polymorphism:
          G/T, Transversion substitution
          Context Sequence [VIC/FAM]:

          TCATGGACCCCACCTTCCTCTTCCA[G/T]TCCATTACCGCCAACATCATCTGCT

          Assay ID C___7817765_C0
          Size 150 rxns
          Availability Inventoried
          Catalog # 4362691
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details



          Species:

          Human

          dbSNP Submissions:

          NA

          Phenotype:

          MIM: 123930

          Literature Links:

          CYP2B6 PubMed Links

          Allele Nomenclature:

          CYP2B6*13A,c.516G>T CYP2B6*13A,g.15631G>T CYP2B6*13B,c.516G>T CYP2B6*13B,g.15631G>T CYP2B6*19,c.516G>T CYP2B6*19,g.15631G>T CYP2B6*20,c.516G>T CYP2B6*20,g.15631G>T CYP2B6*6A,c.516G>T CYP2B6*6A,g.15631G>T CYP2B6*6B,c.516G>T CYP2B6*6B,g.15631G>T CYP2B6*6C,c.516G>T CYP2B6*6C,g.15631G>T CYP2B6*7A,c.516G>T CYP2B6*7A,g.15631G>T CYP2B6*7B,c.516G>T CYP2B6*7B,g.15631G>T CYP2B6*9,c.516G>T CYP2B6*9,g.15631G>T

          Minor Allele Frequency:

          1000Genome Applied Biosystems® HapMap
          Global
          T (0.32)
          (0.68)
          Caucasian
          T (0.17)
          (0.83)
          CEPH (CEU)
          T (0.27)
          (0.73)
          EAS
          T (0.22)
          (0.78)
          Caucasian
          T (0.17)
          (0.83)
          YRI (Yoruba)
          T (0.42)
          (0.58)
          SAS
          T (0.38)
          (0.62)
          African American
          T (0.42)
          (0.58)
          JPT (Japanese)
          T (0.18)
          (0.82)
          AFR
          T (0.37)
          (0.63)
          African American
          T (0.42)
          (0.58)
          CHB (Han Chinese)
          T (0.15)
          (0.85)
          EUR
          T (0.24)
          (0.76)
          Japanese
          T (0.18)
          (0.82)
          AMR
          T (0.37)
          (0.63)
          Japanese
          T (0.18)
          (0.82)
          Chinese
          T (0.30)
          (0.70)
          Chinese
          T (0.30)
          (0.70)
          CYP2B6 - cytochrome P450 family 2 subfamily B member 6
          Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
          NM_000767.4 619 Silent Mutation CAG,CAT Q,H 172 NP_000758.1
          XM_005258569.4 619 Silent Mutation CAG,CAT Q,H 172 XP_005258626.1
          XM_006723050.3 619 Silent Mutation CAG,CAT Q,H 172 XP_006723113.1
          XM_011526546.2 619 Silent Mutation CAG,CAT Q,H 172 XP_011524848.1
          XM_011526547.2 619 Intron XP_011524849.1
          XM_011526548.2 619 Intron XP_011524850.1
          XM_011526549.2 619 Intron XP_011524851.1
          XM_011526550.2 619 Intron XP_011524852.1

          Back To Top

          More Information


          Important Information

          Assay C___7817765_C0 to CYP2B6*6 c.516G>T rs3745274 replaces C___7817765_60. C___7817765_C0 demonstrates superior amplification and cluster separation on OpenArray, and also works well on 96- or 384-well plates when run with Genotyping Master Mix. If using Universal Master Mix, use C___7817765_60 as C___7817765_C0 does not amplify well with Universal Master Mix. C___7817765_60 is discontinued but can still be ordered, if needed.

          Set Membership:

          HapMap DME Validated Inventoried

          Panther Classification:

          Molecular Function -

          oxidoreductase oxygenase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          xenobiotic metabolic process
          steroid metabolic process
          drug metabolic process
          epoxygenase P450 pathway
          cellular ketone metabolic process
          exogenous drug catabolic process
          oxidation-reduction process
          monooxygenase activity
          iron ion binding
          arachidonic acid epoxygenase activity
          steroid hydroxylase activity
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced flavin or flavoprotein as one donor, and incorporation of one atom of oxygen
          oxygen binding
          heme binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline