Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601670 MIM: 616655 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DPF1 - double PHD fingers 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135155.2 | Intron | NP_001128627.1 | ||||
NM_001135156.2 | Intron | NP_001128628.1 | ||||
NM_001289978.1 | Intron | NP_001276907.1 | ||||
NM_004647.3 | Intron | NP_004638.2 | ||||
XM_005259289.2 | Intron | XP_005259346.1 | ||||
XM_005259290.3 | Intron | XP_005259347.1 | ||||
XM_005259292.4 | Intron | XP_005259349.1 | ||||
XM_006723407.2 | Intron | XP_006723470.1 | ||||
XM_006723408.3 | Intron | XP_006723471.1 | ||||
XM_006723409.2 | Intron | XP_006723472.1 | ||||
XM_006723410.2 | Intron | XP_006723473.1 | ||||
XM_011527356.1 | Intron | XP_011525658.1 | ||||
XM_011527357.2 | Intron | XP_011525659.1 | ||||
XM_011527358.2 | Intron | XP_011525660.1 |
SIPA1L3 - signal induced proliferation associated 1 like 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |