Search
Search

Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTPRN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| PTPRN2 - protein tyrosine phosphatase, receptor type N2 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001308267.1 | Intron | NP_001295196.1 | ||||
| NM_001308268.1 | Intron | NP_001295197.1 | ||||
| NM_002847.4 | Intron | NP_002838.2 | ||||
| NM_130842.3 | Intron | NP_570857.2 | ||||
| NM_130843.3 | Intron | NP_570858.2 | ||||
| XM_011516446.1 | Intron | XP_011514748.1 | ||||
| XM_011516447.2 | Intron | XP_011514749.1 | ||||
| XM_011516448.2 | Intron | XP_011514750.1 | ||||
| XM_011516449.2 | Intron | XP_011514751.1 | ||||
| XM_017012475.1 | Intron | XP_016867964.1 | ||||
| XM_017012476.1 | Intron | XP_016867965.1 | ||||