Search
Search

Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C19orf45 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
| C19orf45 - chromosome 19 open reading frame 45 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_198534.2 | 1275 | Silent Mutation | CCC,CCT | P,P 378 | NP_940936.2 | |
| XM_005272473.3 | 1275 | Silent Mutation | CCC,CCT | P,P 378 | XP_005272530.1 | |
| XM_011527993.1 | 1275 | Silent Mutation | CCC,CCT | P,P 353 | XP_011526295.1 | |
| XM_011527995.2 | 1275 | Silent Mutation | CCC,CCT | P,P 378 | XP_011526297.1 | |
| ZNF358 - zinc finger protein 358 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||