Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605741 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR183 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
GPR183 - G protein-coupled receptor 183 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004951.4 | 1721 | UTR 3 | NP_004942.1 | |||
XM_017020405.1 | 1721 | UTR 3 | XP_016875894.1 |
UBAC2 - UBA domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144072.1 | 1721 | Intron | NP_001137544.1 | |||
NM_177967.3 | 1721 | Intron | NP_808882.1 | |||
XM_006719948.3 | 1721 | Intron | XP_006720011.1 | |||
XM_011521082.2 | 1721 | Intron | XP_011519384.1 | |||
XM_011521083.2 | 1721 | Intron | XP_011519385.1 | |||
XM_011521084.2 | 1721 | Intron | XP_011519386.1 | |||
XM_017020553.1 | 1721 | Intron | XP_016876042.1 |
Set Membership: |
HapMap Validated |