Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 243305 MIM: 616717 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
INVS PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
INVS - inversin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318381.1 | 2437 | Intron | NP_001305310.1 | |||
NM_001318382.1 | 2437 | Intron | NP_001305311.1 | |||
NM_014425.4 | 2437 | Intron | NP_055240.2 |
TEX10 - testis expressed 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161584.1 | 2437 | Silent Mutation | TTC,TTT | F,F 895 | NP_001155056.1 | |
NM_017746.3 | 2437 | Silent Mutation | TTC,TTT | F,F 911 | NP_060216.2 | |
XM_011518798.1 | 2437 | Silent Mutation | TTC,TTT | F,F 786 | XP_011517100.1 | |
XM_017014848.1 | 2437 | Silent Mutation | TTC,TTT | F,F 556 | XP_016870337.1 |
Set Membership: |
HapMap Validated |