Search
Search

CTD-2270F17.1
KCNIP1
KCNMB1GTCTCGTGGGAAACAGATTTCAAAC[A/G]ACAACAGACAGCAAGACTGAGAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTD-2270F17.1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||||||||
|---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
| CTD-2270F17.1 - uncharacterized LOC101928033 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| KCNIP1 - potassium voltage-gated channel interacting protein 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001034837.2 | 117 | Intron | NP_001030009.1 | |||
| NM_001034838.2 | 117 | Intron | NP_001030010.1 | |||
| NM_001278339.1 | 117 | Intron | NP_001265268.1 | |||
| NM_001278340.1 | 117 | Intron | NP_001265269.1 | |||
| NM_014592.3 | 117 | Intron | NP_055407.1 | |||
| XM_017009407.1 | 117 | Intron | XP_016864896.1 | |||
| XM_017009408.1 | 117 | Intron | XP_016864897.1 | |||
| KCNMB1 - potassium calcium-activated channel subfamily M regulatory beta subunit 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_004137.3 | 117 | UTR 5 | NP_004128.1 | |||
Set Membership: |
|