Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602335 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EMP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
EMP3 - epithelial membrane protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM143 - transmembrane protein 143 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303538.1 | 1832 | UTR 3 | NP_001290467.1 | |||
NM_001303539.1 | 1832 | UTR 3 | NP_001290468.1 | |||
NM_001303540.1 | 1832 | UTR 3 | NP_001290469.1 | |||
NM_018273.3 | 1832 | UTR 3 | NP_060743.2 |
Set Membership: |
HapMap Validated |