Search
Search

Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
|||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HK1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
| HK1 - hexokinase 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_000188.2 | 519 | Intron | NP_000179.2 | |||
| NM_001322364.1 | 519 | Intron | NP_001309293.1 | |||
| NM_001322365.1 | 519 | Intron | NP_001309294.1 | |||
| NM_001322366.1 | 519 | Intron | NP_001309295.1 | |||
| NM_001322367.1 | 519 | Intron | NP_001309296.1 | |||
| NM_033496.2 | 519 | Intron | NP_277031.1 | |||
| NM_033497.2 | 519 | Intron | NP_277032.1 | |||
| NM_033498.2 | 519 | Intron | NP_277033.1 | |||
| NM_033500.2 | 519 | Missense Mutation | CAT,CGT | H,R 7 | NP_277035.2 | |
| XM_005269737.1 | 519 | Intron | XP_005269794.1 | |||
| XM_011539732.1 | 519 | Missense Mutation | CAT,CGT | H,R 7 | XP_011538034.1 | |
Set Membership: |
|