Search
Search

Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
43 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRIM33 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||||||||
|---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
| TRIM33 - tripartite motif containing 33 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_015906.3 | 4838 | UTR 3 | NP_056990.3 | |||
| NM_033020.2 | 4838 | UTR 3 | NP_148980.2 | |||
| XM_005270936.3 | 4838 | UTR 3 | XP_005270993.1 | |||
| XM_005270937.3 | 4838 | UTR 3 | XP_005270994.1 | |||
| XM_011541568.2 | 4838 | UTR 3 | XP_011539870.1 | |||
| XM_017001452.1 | 4838 | UTR 3 | XP_016856941.1 | |||
| XM_017001453.1 | 4838 | UTR 3 | XP_016856942.1 | |||
| XM_017001454.1 | 4838 | Intron | XP_016856943.1 | |||
Set Membership: |
|