Search
Search

Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IRAK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||||||||
|---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
| IRAK1 - interleukin 1 receptor associated kinase 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001025242.1 | 1437 | Intron | NP_001020413.1 | |||
| NM_001025243.1 | 1437 | Missense Mutation | TCG,TTG | S,L 453 | NP_001020414.1 | |
| NM_001569.3 | 1437 | Missense Mutation | TCG,TTG | S,L 532 | NP_001560.2 | |
| XM_005274668.3 | 1437 | Intron | XP_005274725.1 | |||
| XM_011531158.2 | 1437 | Intron | XP_011529460.1 | |||
| MECP2 - methyl-CpG binding protein 2 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| MIR718 - microRNA 718 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||