Search
Search

Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PEMT PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||||||||
|---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
| PEMT - phosphatidylethanolamine N-methyltransferase | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001267551.1 | 498 | Missense Mutation | ATG,GTG | M,V 190 | NP_001254480.1 | |
| NM_001267552.1 | 498 | Missense Mutation | AAT,AGT | N,S 222 | NP_001254481.1 | |
| NM_007169.2 | 498 | Missense Mutation | ATG,GTG | M,V 175 | NP_009100.2 | |
| NM_148172.2 | 498 | Missense Mutation | ATG,GTG | M,V 212 | NP_680477.1 | |
| NM_148173.1 | 498 | Missense Mutation | ATG,GTG | M,V 175 | NP_680478.1 | |
| XM_006721418.3 | 498 | Missense Mutation | ATG,GTG | M,V 191 | XP_006721481.2 | |
| XM_017024016.1 | 498 | Missense Mutation | ATG,GTG | M,V 101 | XP_016879505.1 | |
| RASD1 - ras related dexamethasone induced 1 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
Set Membership: |
|