Search
Search

Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCA7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
| ABCA7 - ATP binding cassette subfamily A member 7 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| CNN2 - calponin 2 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001303499.1 | 771 | Intron | NP_001290428.1 | |||
| NM_001303501.1 | 771 | Silent Mutation | GCG,GCT | A,A 128 | NP_001290430.1 | |
| NM_004368.3 | 771 | Silent Mutation | GCG,GCT | A,A 128 | NP_004359.1 | |
| NM_201277.2 | 771 | Silent Mutation | GCG,GCT | A,A 128 | NP_958434.1 | |