Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603950 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NDST3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NDST3 - N-deacetylase and N-sulfotransferase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004784.2 | Intron | NP_004775.1 | ||||
XM_006714416.3 | Intron | XP_006714479.1 | ||||
XM_011532415.2 | Intron | XP_011530717.1 | ||||
XM_011532416.2 | Intron | XP_011530718.1 | ||||
XM_017008839.1 | Intron | XP_016864328.1 | ||||
XM_017008840.1 | Intron | XP_016864329.1 | ||||
XM_017008841.1 | Intron | XP_016864330.1 | ||||
XM_017008842.1 | Intron | XP_016864331.1 | ||||
XM_017008843.1 | Intron | XP_016864332.1 | ||||
XM_017008844.1 | Intron | XP_016864333.1 |
Set Membership: |
Validated |