Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
AGCAATTGCTATGCTAAAGGGTAGT[G/T]CCAAATTTAGCAAATACCAGCTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607724 MIM: 610394 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAPS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CAPS2 - calcyphosine 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GLIPR1L2 - GLI pathogenesis-related 1 like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270396.1 | Intron | NP_001257325.1 | ||||
NM_152436.2 | Intron | NP_689649.1 | ||||
XM_011537949.2 | Intron | XP_011536251.1 | ||||
XM_011537950.2 | Intron | XP_011536252.1 | ||||
XM_017018848.1 | Intron | XP_016874337.1 | ||||
XM_017018849.1 | Intron | XP_016874338.1 |
Set Membership: |
Validated |