Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
CATGTTTCATTTCTGTGCCAACATA[C/T]AGGAGACTATTGCAGGTGGCATTTA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 605497 | |||||||||||||||||||||||
Literature Links: |
CRTAP PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CRTAP - cartilage associated protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SUSD5 - sushi domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015551.1 | 4062 | UTR 3 | NP_056366.1 | |||
XM_005265034.3 | 4062 | UTR 3 | XP_005265091.1 | |||
XM_017006137.1 | 4062 | UTR 3 | XP_016861626.1 |