Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCCAACAAAGGTTTGTTGAGTCC[C/G]TATTACGTATAAACAGTGTGCCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
37 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606168 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDX20 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DDX20 - DEAD-box helicase 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007204.4 | Intron | NP_009135.4 |
FAM212B - family with sequence similarity 212 member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FAM212B-AS1 - FAM212B antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC101928718 - uncharacterized LOC101928718 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |