Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603122 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DOCK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DOCK2 - dedicator of cytokinesis 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004946.2 | 2901 | Intron | NP_004937.1 | |||
XM_005265830.3 | 2901 | Intron | XP_005265887.1 | |||
XM_011534448.2 | 2901 | Intron | XP_011532750.1 | |||
XM_011534449.2 | 2901 | Intron | XP_011532751.1 | |||
XM_011534450.2 | 2901 | Intron | XP_011532752.1 | |||
XM_011534451.2 | 2901 | Intron | XP_011532753.1 | |||
XM_017009189.1 | 2901 | Missense Mutation | CCC,GCC | P,A 941 | XP_016864678.1 | |
XM_017009190.1 | 2901 | Intron | XP_016864679.1 |
FAM196B - family with sequence similarity 196 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001129891.1 | 2901 | Intron | NP_001123363.1 | |||
XM_011534415.2 | 2901 | Intron | XP_011532717.1 | |||
XM_017008908.1 | 2901 | Intron | XP_016864397.1 |
Set Membership: |
HapMap |