Search
Search

LOC103344931
VTI1AGAGAACCCTCATGTCTAGGGAGCAC[C/T]CCATGGGGTCATCTCCATGCCTGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC103344931 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
| LOC103344931 - uncharacterized LOC103344931 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| VTI1A - vesicle transport through interaction with t-SNAREs 1A | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001318203.1 | 11410 | Intron | NP_001305132.1 | |||
| NM_001318205.1 | 11410 | Intron | NP_001305134.1 | |||
| NM_145206.3 | 11410 | Intron | NP_660207.2 | |||
| XM_005269544.4 | 11410 | Intron | XP_005269601.1 | |||
| XM_005269545.4 | 11410 | Intron | XP_005269602.1 | |||
| XM_005269546.3 | 11410 | UTR 3 | XP_005269603.1 | |||
| XM_005269547.4 | 11410 | Intron | XP_005269604.1 | |||
| XM_006717637.2 | 11410 | Intron | XP_006717700.1 | |||
| XM_011539328.2 | 11410 | Intron | XP_011537630.1 | |||
| XM_011539330.2 | 11410 | Intron | XP_011537632.1 | |||
| XM_011539331.2 | 11410 | Intron | XP_011537633.1 | |||
| XM_011539333.2 | 11410 | Intron | XP_011537635.1 | |||
| XM_017015745.1 | 11410 | Intron | XP_016871234.1 | |||
| XM_017015746.1 | 11410 | Intron | XP_016871235.1 | |||
| XM_017015747.1 | 11410 | Intron | XP_016871236.1 | |||
| XM_017015748.1 | 11410 | Intron | XP_016871237.1 | |||
Set Membership: |
|