Search
Search

Notch1GGGTGGACAATTTTCACTTCTCCCT[A/G]CTCTGAAACAGCATGTCTCCACACA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Notch1 PubMed Links |
| Notch1 - notch 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_008714.3 | Intron | NP_032740.3 | ||||