Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTATAAGTAAGTGTTCCTCATTTCT[G/A]AGACTTTCCCGATAGTTACCTCCAC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Fbxo5 PubMed Links |
Fbxo5 - F-box protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025995.2 | Intron | NP_080271.2 |