Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Klf7 PubMed Links |
Klf7 - Kruppel-like factor 7 (ubiquitous) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033563.2 | Intron | NP_291041.2 |
Mir6899 - microRNA 6899 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |