Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCAAGCTGGCTTTTCGCCCAGCAC[A/G]GCAGCTCTCAATCAGACACCGTGAC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Csnk1d PubMed Links |
Csnk1d - casein kinase 1, delta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_027874.2 | Intron | NP_082150.1 | ||||
NM_139059.2 | Intron | NP_620690.1 |