Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
ACAAAAAGAAAGAAGAGAAGGAAGA[C/T]GAAATCTCATTAGAAGATCTAATTG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Zc3h15 PubMed Links |
Zc3h15 - zinc finger CCCH-type containing 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_026934.3 | 759 | Silent Mutation | NP_081210.2 |