Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
GCCACCATACCATTAGAACTACAGA[A/G]CACTTTCTCCCCAAGCCATACCTAT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Cep112 PubMed Links |
Cep112 - centrosomal protein 112 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_029586.2 | Intron | NP_083862.1 | ||||
NM_029606.3 | Intron | NP_083882.3 | ||||
NM_145688.2 | Intron | NP_663721.2 |