Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
GACTTTCTTTGAAATAGTCAGGTGC[A/G]CACCAGTTGGAGGGCTGGCGGAAAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Fbn2 PubMed Links |
Fbn2 - fibrillin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_010181.2 | 9470 | UTR 3 | NP_034311.2 |